N(α)-Benzyloxycarbonyl-L-lysine 98

Supplier: Thermo Fisher Scientific

Synonyms: (S)-6-Amino-2-(((benzyloxy)carbonyl)amino)hexanoic acid, Z-Lys-OH

L08629.03 L08629.06
ALFAL08629.03EA 77.9 SGD
ALFAL08629.03 ALFAL08629.06
N(α)-Benzyloxycarbonyl-L-lysine 98
N(α)-Benzyloxycarbonyl-L-lysine

Purity: 98+% CAS No.: 2212-75-1 Molecular Formula: C14H20N2O4 Formula Weight: 280.32 Storage Temperature: 2°C to 8°C Melting Point: ca 237° dec. Beilstein Reference: 2153826 Optical Rotation: -12° (c=2 in 0.2N HCl) MDL No.: MFCD00038204

Formula: C₁₄H₂₀N₂O₄
MW: 280.3 g/mol
Melting Pt: ∼ 237 °C (dec.)
MDL Number: MFCD00038204
CAS Number: 2212-75-1
EINECS: 218-662-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR