4-Bromo-2-methoxyphenol ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-Bromoguaiacol, 5-Brom-2-hydroxyanisole

L08801.09 L08801.18 L08801.04
ALFAL08801.09EA 294 SGD
ALFAL08801.09 ALFAL08801.18 ALFAL08801.04
4-Bromo-2-methoxyphenol ≥98%
4-Bromo-2-methoxyphenol

Purity: 98% CAS No.: 7368-78-7 Molecular Formula: C7H7BrO2 Formula Weight: 203.04 Odour: Phenol-like Melting Point: 32-37° Boiling Point: 129-132°/12mm Beilstein Reference: 2045286 MDL No.: MFCD00051937

Formula: C₇H₇BrO₂
Boiling Pt: 129…132 °C (12 mmHg)
Melting Pt: 32…37 °C
Storage Temperature: Ambient
MDL Number: MFCD00051937
CAS Number: 7368-78-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR