6-Methylnicotinic acid 99

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 6-Methyl-3-pyridinecarboxylic acid, 6-Methylpyridine-3-carboxylic acid

L08877.03 L08877.06 L08877.14
ALFAL08877.03EA 96.9 SGD
ALFAL08877.03 ALFAL08877.06 ALFAL08877.14
6-Methylnicotinic acid 99
6-Methylnicotinic acid

Purity: 99% CAS No.: 3222-47-7 Molecular Formula: C7H7NO2 Formula Weight: 137.14 Melting Point: 210-215° Beilstein Reference: 112049

Formula: C₇H₇NO₂
MW: 137.14 g/mol
Melting Pt: 210…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00006341
CAS Number: 3222-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR