2-Chlorobenzenesulphonamide ≥98%

Supplier: Thermo Fisher Scientific

Synonyms: o-Chlorobenzenesulphonamide

L09082.03 L09082.06
ALFAL09082.03EA 59.5 SGD
ALFAL09082.03 ALFAL09082.06
2-Chlorobenzenesulphonamide ≥98%
2-Chlorobenzenesulphonamide

Purity: 98% CAS No.: 6961-82-6 Molecular Formula: C6H6ClNO2S Formula Weight: 191.64 Melting Point: 189-191° Beilstein Reference: 2937540

Formula: C₆H₆ClNO₂S
MW: 191.64 g/mol
Melting Pt: 189…191 °C
Storage Temperature: Ambient
MDL Number: MFCD00051974
CAS Number: 6961-82-6
EINECS: 230-156-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR