(S)-Methyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride 95%

Supplier: Thermo Fisher Scientific

Synonyms: H-Lys(Z)-OMe.HCl, H-Lys(Z)-OMe·HCl

L09744.03 L09744.06
ALFAL09744.03EA 85.2 SGD
ALFAL09744.03 ALFAL09744.06
(S)-Methyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride 95%
(S)-Methyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride

Purity: 95% CAS No.: 27894-50-4 Molecular Formula: C15H22N2O4.HCl Formula Weight: 330.81 Storage Temperature: 2°C to 8°C Melting Point: 115-118° Beilstein Reference: 3578476 Optical Rotation: +14.5° (c=2 in water) MDL No.: MFCD00034846

Formula: C₁₅H₂₃ClN₂O₄
MW: 330.809 g/mol
Melting Pt: 115…118 °C
MDL Number: MFCD00034846
CAS Number: 27894-50-4
EINECS: 248-715-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR