(S)-(+)-Prolinol ≥98%

Supplier: Thermo Fisher Scientific
Warning

L09779.03 L09779.06 L09779.14
ALFAL09779.03EA 75.7 SGD
ALFAL09779.03 ALFAL09779.06 ALFAL09779.14
(S)-(+)-Prolinol ≥98%
(S)-(+)-Prolinol

Purity: 98% CAS No.: 23356-96-9 Molecular Formula: C5H11NO Formula Weight: 101.15 Flash Point: 86°(186°F) Refractive Index: 1.4853 Melting Point: 42-44° Boiling Point: 74-76°/2mm Beilstein Reference: 79843 Optical Rotation: +31° (c=1 in toluene) MDL No.: MFCD00005255

Formula: C₅H₁₁NO
Boiling Pt: 74…76 °C (2 mmHg)
Melting Pt: 42…44 °C
Density: 1,025
Flash Pt: 86 °C (186 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005255
CAS Number: 23356-96-9
EINECS: 245-605-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR