n-Hexane 99

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

L09938.AD L09938.AK L09938.AU
ALFAL09938.ADEA 63 SGD
ALFAL09938.AD ALFAL09938.AK ALFAL09938.AU
n-Hexane 99
n-Hexane

Purity: 99% CAS No.: 110-54-3 Molecular Formula: CH3(CH2)4CH3 Formula Weight: 86.18 Flash Point: -23°(-9°F) Refractive Index: 1.3750 Odour: Weak, characteristic Melting Point: -95° Boiling Point: 69° Beilstein Reference: 1730733 Merck Reference: 14,4694 UN No.: 1208 MDL No.: MFCD00009520

Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69 °C
Melting Pt: –95 °C
Density: 0.659
Flash Pt: –23 °C (–9 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 14,4694
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR