Bis(2-ethylhexyl)adipate 99

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Di-(2-ethylhexyl) adipate, Bis(2-ethylhexyl) hexanedioate, Dioctyl Adipate, Diisooctyl adipate, Bis(2-ethylhexyl) adipate

L10195.14 L10195.22
ALFAL10195.14EA 71.5 SGD
ALFAL10195.14 ALFAL10195.22
Bis(2-ethylhexyl)adipate 99
Bis(2-ethylhexyl) adipate

Purity: 99% CAS No.: 103-23-1 Molecular Formula: C22H42O4 Formula Weight: 370.58 Flash Point: 196°(384°F) Refractive Index: 1.4470 Melting Point: -67° Boiling Point: 166-168°/1mm Beilstein Reference: 1803774 MDL No.: MFCD00009496

Formula: C₂₂H₄₂O₄
MW: 370.57 g/mol
Boiling Pt: 166…168 °C (1 mmHg)
Melting Pt: –67 °C
Density: 0.990
Flash Pt: 196 °C (384 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009496
CAS Number: 103-23-1
EINECS: 203-090-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR