Chlorophenol red sodium salt

Supplier: Thermo Fisher Scientific

L10364.06
ALFAL10364.06EA 150 SGD
ALFAL10364.06
Chlorophenol red sodium salt
Chlorophenol red sodium salt

CAS No.: 123333-64-2 Molecular Formula: C19H11Cl2NaO5S Formula Weight: 445.26 Melting Point: ca 267° dec. MDL No.: MFCD00151199

Formula: C₁₉H₁₂Cl₂O₅S
Melting Pt: ∼ 267 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00151199
CAS Number: 123333-64-2
EINECS: 224-619-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR