5-Nitro-2-furonitrile ≥97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 5-Nitrofuran-2-carbonitrile, 2-Cyano-5-nitrofuran

L10486.06 L10486.14
ALFAL10486.06EA 191 SGD
ALFAL10486.06 ALFAL10486.14
5-Nitro-2-furonitrile ≥97%
5-Nitro-2-furonitrile

Purity: 97% CAS No.: 59-82-5 Molecular Formula: C5H2N2O3 Formula Weight: 138.08 Melting Point: 61-64° Beilstein Reference: 123933 UN No.: 3439 MDL No.: MFCD00082647

Formula: C₅H₂N₂O₃
MW: 138.08 g/mol
Melting Pt: 61…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00082647
CAS Number: 59-82-5
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR