(R)-(-)-Prolinol ≥98%

Supplier: Thermo Fisher Scientific
Warning

L11109.03 L11109.MD L11109.06 L11109.14
ALFAL11109.03EA 57.5 SGD
ALFAL11109.03 ALFAL11109.MD ALFAL11109.06 ALFAL11109.14
(R)-(-)-Prolinol ≥98%
(R)-(-)-Prolinol

Purity: 98+% CAS No.: 68832-13-3 Molecular Formula: C5H11NO Formula Weight: 101.15 Flash Point: 86°(186°F) Refractive Index: 1.4849 Boiling Point: 74-76°/2mm Beilstein Reference: 1523669 Optical Rotation: -31° (c=1 in toluene) MDL No.: MFCD00064321

Formula: C₅H₁₁NO
Boiling Pt: 74…76 °C (2 mmHg)
Density: 1,025
Flash Pt: 86 °C (186 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064321
CAS Number: 68832-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR