Methyl-2-amino-4-chlorobenzoate 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Methyl 2-amino-4-chlorobenzoate

L11125.09 L11125.18
ALFAL11125.09EA 155 SGD
ALFAL11125.09 ALFAL11125.18
Methyl-2-amino-4-chlorobenzoate 98%
Methyl 2-amino-4-chlorobenzoate

Purity: 98% CAS No.: 5900-58-3 Molecular Formula: C8H8ClNO2 Formula Weight: 185.61 Melting Point: 66-68° Beilstein Reference: 1072895

Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
Melting Pt: 66…68 °C
Storage Temperature: Ambient
MDL Number: MFCD00017568
CAS Number: 5900-58-3
EINECS: 227-593-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR