Ethyl-4-(trifluoromethyl)benzoate ≥99%

Supplier: Thermo Fisher Scientific

Synonyms: Ethyl 4-(trifluoromethyl)benzoate

L11139.03
ALFAL11139.03EA 83.4 SGD
ALFAL11139.03
Ethyl-4-(trifluoromethyl)benzoate ≥99%
Ethyl 4-(trifluoromethyl)benzoate

Purity: 99% CAS No.: 583-02-8 Molecular Formula: C10H9F3O2 Formula Weight: 218.18 Refractive Index: 1.4470 Boiling Point: 78-80°/5mm MDL No.: MFCD00013563

Formula: C₁₀H₉F₃O₂
MW: 218.18 g/mol
Boiling Pt: 78…80 °C (5 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00013563
CAS Number: 583-02-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR