Sodium-4-aminoazobenzene-4'-sulphonate 90%, tech.

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-Aminoazobenzene-4'-sulphonic acid Sodium Salt

L11965.14 L11965.22
ALFAL11965.14EA 81.8 SGD
ALFAL11965.14 ALFAL11965.22
Sodium-4-aminoazobenzene-4'-sulphonate 90%, tech.
Sodium-4-aminoazobenzene-4'-sulphonate

Purity: 90% CAS No.: 2491-71-6 Molecular Formula: C12H10N3NaO3S Formula Weight: 299.29 Beilstein Reference: 8098548 MDL No.: MFCD00035564

Formula: C₁₂H₁₁N₃O₃S
MW: 299.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00035564
CAS Number: 2491-71-6
EINECS: 219-657-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR