2-(4-Morpholino)ethyl isothiocyanate ≥97%

Supplier: Thermo Fisher Scientific
Danger

L12046.04
ALFAL12046.04EA 183 SGD
ALFAL12046.04
2-(4-Morpholino)ethyl isothiocyanate ≥97%
2-(4-Morpholino)ethyl isothiocyanate

Purity: 97% CAS No.: 63224-35-1 Molecular Formula: C7H12N2OS Formula Weight: 172.25 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.5390 Boiling Point: 162-163°/10mm Beilstein Reference: 1210530 UN No.: 2922 MDL No.: MFCD00041221

Formula: C₇H₁₂N₂OS
MW: 172.25 g/mol
Boiling Pt: 162…163 °C (10 mmHg)
Density: 1.110
Flash Pt: >110 °C (230 °F)
MDL Number: MFCD00041221
CAS Number: 63224-35-1
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR