4-Amino-3-chloro-5-methylbenzonitrile tech. 90%, Technical Grade

Supplier: Thermo Fisher Scientific
Danger

L12541.03
ALFAL12541.03EA 0 SGD
ALFAL12541.03
4-Amino-3-chloro-5-methylbenzonitrile tech. 90%, Technical Grade
4-Amino-3-chloro-5-methylbenzonitrile

Purity: 90% CAS No.: 158296-69-6 Molecular Formula: C8H7ClN2 Formula Weight: 166.61 Melting Point: 128-130° Beilstein Reference: 7527075 UN No.: 3439 MDL No.: MFCD00041476

Formula: C₈H₇ClN₂
Melting Pt: 128…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00041476
CAS Number: 158296-69-6
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR