Methyl-4-bromobutyrate 97

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Methyl 4-bromobutanoate

L12636.09
ALFAL12636.09EA 93 SGD
ALFAL12636.09
Methyl-4-bromobutyrate 97
Methyl 4-bromobutyrate

Purity: 97% CAS No.: 4897-84-1 Molecular Formula: Br(CH2)3CO2CH3 Formula Weight: 181.04 Refractive Index: 1.4620 Boiling Point: 186-187° MDL No.: MFCD00041482

Formula: C₅H₉BrO₂
MW: 181.03 g/mol
Boiling Pt: 186…187 °C
Storage Temperature: Ambient
MDL Number: MFCD00041482
CAS Number: 4897-84-1
EINECS: 225-523-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR