2-Acetylbenzofuran ≥99%

Supplier: Thermo Fisher Scientific

L13771.06 L13771.14
ALFAL13771.06EA 98.1 SGD
ALFAL13771.06 ALFAL13771.14
2-Acetylbenzofuran ≥99%
2-Acetylbenzofuran

Purity: 99% CAS No.: 1646-26-0 Molecular Formula: C10H8O2 Formula Weight: 160.17 Flash Point: 110°(230°F) Melting Point: 70-72° Boiling Point: 110-113°/3mm Beilstein Reference: 117910 MDL No.: MFCD00005849

Formula: C₁₀H₈O₂
Boiling Pt: 110…113 °C (3 mmHg)
Melting Pt: 70…72 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005849
CAS Number: 1646-26-0
EINECS: 216-706-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR