4-Bromo-3-chlorophenol ≥98%

Supplier: Thermo Fisher Scientific
Warning

L13801.03 L13801.06
ALFAL13801.03EA 0 SGD
ALFAL13801.03 ALFAL13801.06
4-Bromo-3-chlorophenol ≥98%
4-Bromo-3-chlorophenol

Purity: 98% CAS No.: 13631-21-5 Molecular Formula: C6H4BrClO Formula Weight: 207.46 Odour: Phenol-like Melting Point: 64-68° Beilstein Reference: 3236445 MDL No.: MFCD00070744

Formula: C₆H₄BrClO
MW: 207.45 g/mol
Melting Pt: 64…68 °C
Storage Temperature: Ambient
MDL Number: MFCD00070744
CAS Number: 13631-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR