4-Hydroxyindole ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: Indol-4-ol, 4-Indolol, 4-Hydroxy-1H-indole, 1H-Indol-4-ol

ALFAL14352.MEEA 0 SGD
ALFAL14352.ME
4-Hydroxyindole ≥98%
4-Hydroxyindole

Purity: 98% CAS No.: 2380-94-1 Molecular Formula: C8H7NO Formula Weight: 133.15 Melting Point: 97-99° Beilstein Reference: 114905

Formula: C₈H₇NO
MW: 133,15 g/mol
Boiling Pt: 100 °C (0,01 mmHg)
Melting Pt: 97…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00005667
CAS Number: 2380-94-1
EINECS: 219-177-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR