S-Trityl-L-cysteine 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: H-Cys(Trt)-OH

L14384.03 L14384.06
ALFAL14384.03EA 81.3 SGD
ALFAL14384.03 ALFAL14384.06
S-Trityl-L-cysteine 97%
S-Trityl-L-cysteine

Purity: 97% CAS No.: 2799-07-7 Molecular Formula: C22H21NO2S Formula Weight: 363.48 Melting Point: ca 185° dec. Beilstein Reference: 2339626 Optical Rotation: +84° (c=1 in acetic acid) MDL No.: MFCD00002611

Formula: C₂₂H₂₁NO₂S
MW: 363.5 g/mol
Melting Pt: ∼ 185 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00002611
CAS Number: 2799-07-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR