1-Bromo-6-phenylhexane 97%

Supplier: Thermo Fisher Scientific

L14568.03 L14568.06 L14568.MD
ALFAL14568.03EA 271 SGD
ALFAL14568.03 ALFAL14568.06 ALFAL14568.MD
1-Bromo-6-phenylhexane 97%
1-Bromo-6-phenylhexane

Purity: 97% CAS No.: 27976-27-8 Molecular Formula: C6H5(CH2)6Br Formula Weight: 241.18 Refractive Index: 1.5295 Boiling Point: 115-118°/6mm Beilstein Reference: 2207649 MDL No.: MFCD00236022

Formula: C₆H₅(CH₂)₆Br
Boiling Pt: 115…118 °C (6 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00236022
CAS Number: 27976-27-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR