4-Bromo-3-methylbenzamide 99

Supplier: Thermo Fisher Scientific

L16176.14 L16176.03 L16176.06
ALFAL16176.14EA 422 SGD
ALFAL16176.14 ALFAL16176.03 ALFAL16176.06
4-Bromo-3-methylbenzamide 99
4-Bromo-3-methylbenzamide

Purity: 99% CAS No.: 170229-98-8 Molecular Formula: C8H8BrNO Formula Weight: 214.06 Melting Point: 177-182° Beilstein Reference: 868074

Formula: C₈H₈BrNO
MW: 214.06 g/mol
Melting Pt: 177…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00672914
CAS Number: 170229-98-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR