5-Methylnicotinic acid 97

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 5-Methyl-3-pyridinecarboxylic acid, 5-Methylpyridine-3-carboxylic acid

L16370.03 L16370.06
ALFAL16370.03EA 251 SGD
ALFAL16370.03 ALFAL16370.06
5-Methylnicotinic acid 97
5-Methylnicotinic acid

Purity: 97% CAS No.: 3222-49-9 Molecular Formula: C7H7NO2 Formula Weight: 137.14 Melting Point: 210-215° Beilstein Reference: 112288

Formula: C₇H₇NO₂
MW: 137.14 g/mol
Melting Pt: 210…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00829036
CAS Number: 3222-49-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR