Methyl-5-methylnicotinate 94

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Methyl 5-methylnicotinate

L16371.03 L16371.06
ALFAL16371.03EA 110 SGD
ALFAL16371.03 ALFAL16371.06
Methyl-5-methylnicotinate 94
Methyl 5-methylnicotinate

Purity: 94% CAS No.: 29681-45-6 Molecular Formula: C8H9NO2 Formula Weight: 151.17 Melting Point: 42-46° Boiling Point: 90°/3mm MDL No.: MFCD01075739

Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 90 °C (3 mmHg)
Melting Pt: 42…46 °C
Storage Temperature: Ambient
MDL Number: MFCD01075739
CAS Number: 29681-45-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR