Ethyl dibromofluoroacetate ≥97%

Supplier: Thermo Fisher Scientific
Danger

L16491.06 L16491.14
ALFAL16491.06EA 244 SGD
ALFAL16491.06 ALFAL16491.14
Ethyl dibromofluoroacetate ≥97%
Ethyl dibromofluoroacetate

Purity: 97% CAS No.: 565-53-7 Molecular Formula: Br2C4FH5O2 Formula Weight: 263.89 Flash Point: 110°(230°F) Refractive Index: 1.4635 Boiling Point: 82°/33mm Beilstein Reference: 1762048 UN No.: 2922 MDL No.: MFCD00042068

Formula: C₄H₅Br₂FO₂
Boiling Pt: 82 °C (33 mmHg)
Density: 1,894
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042068
CAS Number: 565-53-7
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR