Ultramark1621 Mass Spec Std

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Massmark 1621

L16698.03 L16698.MD
ALFAL16698.03EA 290 SGD
ALFAL16698.03 ALFAL16698.MD
Ultramark1621 Mass Spec Std
Ultramark® 1621

CAS No.: 105809-15-2 Formula Weight: 2120.00 Refractive Index: 1.3530 MDL No.: MFCD00242454

Formula: C₆H₁₈N₃P₃
MW: 231.189 g/mol
Density: 1.8
Storage Temperature: Ambient
MDL Number: MFCD00242454
CAS Number: 105809-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR