2-Bromo-4,5-difluorophenol ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-Bromo-3,4-difluoro-6-hydroxybenzene

L16716.06
ALFAL16716.06EA 227 SGD
ALFAL16716.06
2-Bromo-4,5-difluorophenol ≥97%
2-Bromo-4,5-difluorophenol

Purity: 97% CAS No.: 166281-37-4 Molecular Formula: C6H3BrF2O Formula Weight: 208.99 Refractive Index: 1.5320 Beilstein Reference: 8355005

Formula: C₆H₃BrF₂O
MW: 208.99 g/mol
Density: 1.829
Storage Temperature: Ambient
MDL Number: MFCD00070751
CAS Number: 166281-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR