3-Bromo-N,N-dimethylaniline ≥97%

Supplier: Thermo Fisher Scientific
Danger

L17279.06 L17279.14
ALFAL17279.06EA 179 SGD
ALFAL17279.06 ALFAL17279.14
3-Bromo-N,N-dimethylaniline ≥97%
3-Bromo-N,N-dimethylaniline

Purity: 97% CAS No.: 16518-62-0 Molecular Formula: C8H10BrN Formula Weight: 200.08 Refractive Index: 1.6015 Melting Point: 11° Boiling Point: 126°/14mm Beilstein Reference: 2079908 MDL No.: MFCD00045020

Formula: C₈H₁₀BrN
MW: 200.08 g/mol
Boiling Pt: 126 °C (14 mmHg)
Melting Pt: 11
Density: 1.365
Storage Temperature: Ambient
MDL Number: MFCD00045020
CAS Number: 16518-62-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR