N-(2-Pyridyl)bis(trifluoromethanesulphonimide) 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: N,N-Bis(trifluoromethylsulphonyl)-2-pyridylamine, 2-Pyridyltriflimide, 2-[N,N-Bis(trifluoromethylsulphonyl)amino]pyridine, 1,1,1-Trifluoro-N-(pyridin-2-yl)-N-((trifluoromethyl)sulphonyl)methanesulphonamide

L17433.03 L17433.06
ALFAL17433.03EA 0 SGD
ALFAL17433.03 ALFAL17433.06
N-(2-Pyridyl)bis(trifluoromethanesulphonimide) 98%
N-(2-Pyridyl)bis(trifluoromethanesulphonimide)

Purity: 98% CAS No.: 145100-50-1 Molecular Formula: C7H4F6N2O4S2 Formula Weight: 358.24 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 37-41° Boiling Point: 97°/2mm Beilstein Reference: 5832565 MDL No.: MFCD00191834

Formula: C₇H₄F₆N₂O₄S₂
MW: 358.23 g/mol
Boiling Pt: 97 °C (2 mmHg)
Melting Pt: 37…41 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00191834
CAS Number: 145100-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR