4-Bromo-m-cresol 98

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-Bromo-4-hydroxy-2-methylbenzene, 4-Bromo-3-methylphenol

L17510.03 L17510.06
ALFAL17510.03EA 71.4 SGD
ALFAL17510.03 ALFAL17510.06
4-Bromo-m-cresol 98
4-Bromo-m-cresol

Purity: 98% CAS No.: 14472-14-1 Molecular Formula: C7H7BrO Formula Weight: 187.04 Odour: Phenol-like Melting Point: 57-61° Boiling Point: 142-145°/23mm Beilstein Reference: 1859122 MDL No.: MFCD00079723

Formula: C₇H₇BrO
MW: 187.04 g/mol
Boiling Pt: 142…145 °C (23 mmHg)
Melting Pt: 57…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00079723
CAS Number: 14472-14-1
EINECS: 238-464-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR