3-Chlor-1,2-benzisothiazol ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 3-Chlorobenzo[d]isothiazole, 3-Chloro-1,2-benzisothiazole

L17791.06 L17791.14
ALFAL17791.06EA 187 SGD
ALFAL17791.06 ALFAL17791.14
3-Chlor-1,2-benzisothiazol ≥97%
3-Chlor-1,2-benzisothiazol

Purity: 99% CAS No.: 7716-66-7 Molecular Formula: C7H4ClNS Formula Weight: 169.63 Melting Point: 37-44° Boiling Point: 80-86°/0.75mm MDL No.: MFCD00673254

Formula: C₇H₄ClNS
MW: 169.63 g/mol
Boiling Pt: 80…86 °C (0.75 mmHg)
Melting Pt: 37…44 °C
Storage Temperature: Ambient
MDL Number: MFCD00673254
CAS Number: 7716-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR