Ethyl-4-acetylbutyrate 97%

Supplier: Thermo Fisher Scientific
Warning

L17848.22 L17848.06 L17848.14
ALFAL17848.22EA 1010 SGD
ALFAL17848.22 ALFAL17848.06 ALFAL17848.14
Ethyl-4-acetylbutyrate 97%
Ethyl-4-acetylbutyrate

Purity: 97% CAS No.: 13984-57-1 Molecular Formula: C8H14O3 Formula Weight: 158.20 Flash Point: 69°(156°F) Refractive Index: 1.4275 Boiling Point: 65°/0.76mm Beilstein Reference: 1762625 MDL No.: MFCD00009213

Formula: C₈H₁₄O₃
Boiling Pt: 65 °C (0,76 mmHg)
Density: 0,989
Flash Pt: 69 °C (156 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009213
CAS Number: 13984-57-1
EINECS: 237-776-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR