3-Fluoro-4-formylphenylboronic acid ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 3-Fluoro-4-formylbenzeneboronic acid

L17851.03
ALFAL17851.03EA 116 SGD
ALFAL17851.03
3-Fluoro-4-formylphenylboronic acid ≥98%
3-Fluoro-4-formylphenylboronic acid

Purity: 98% CAS No.: 248270-25-9 Molecular Formula: C7H6BFO3 Formula Weight: 167.93 Melting Point: ca 240° MDL No.: MFCD02093051

Formula: C₇H₆BFO₃
MW: 167,93 g/mol
Melting Pt: ∼ 240 °C
Storage Temperature: Ambient
MDL Number: MFCD02093051
CAS Number: 248270-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR