1-Bromo-2-cyclohexylbenzene ≥97%

Supplier: Thermo Fisher Scientific

L18059.14 L18059.06 L18059.03
ALFAL18059.14EA 1220 SGD
ALFAL18059.14 ALFAL18059.06 ALFAL18059.03
1-Bromo-2-cyclohexylbenzene ≥97%
1-Bromo-2-cyclohexylbenzene

Purity: 97% CAS No.: 59734-92-8 Molecular Formula: C12H15Br Formula Weight: 239.16 Refractive Index: 1.5605 Boiling Point: 79-83°/0.01mm MDL No.: MFCD02093987

Formula: C₁₂H₁₅Br
MW: 239.16 g/mol
Boiling Pt: 79…83 °C (0.01 mmHg)
Density: 1.295
Storage Temperature: Ambient
MDL Number: MFCD02093987
CAS Number: 59734-92-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR