5-Bromo-2-chloropyrimidine ≥98%

Supplier: Thermo Fisher Scientific
Warning

L18360.06 L18360.03 L18360.14
ALFAL18360.06EA 286 SGD
ALFAL18360.06 ALFAL18360.03 ALFAL18360.14
5-Bromo-2-chloropyrimidine ≥98%
5-Bromo-2-chloropyrimidine

Purity: 98+% CAS No.: 32779-36-5 Molecular Formula: C4H2BrClN2 Formula Weight: 193.43 Melting Point: 77-80° Boiling Point: 95°/15mm MDL No.: MFCD00483232

Formula: C₄H₂BrClN₂
Boiling Pt: 95 °C (15 mmHg)
Melting Pt: 77…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00483232
CAS Number: 32779-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR