Bis(neopentyl glycolato)diboron 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 5,5,5',5'-Tetramethyl-2,2'-bi(1,3,2-dioxaborinane), 2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)-5,5-dimethyl-1,3,2-dioxaborinane

L18675.06 L18675.03 L18675.MD
ALFAL18675.06EA 0 SGD
ALFAL18675.06 ALFAL18675.03 ALFAL18675.MD
Bis(neopentyl glycolato)diboron 97%
Bis(neopentyl glycolato)diboron

Purity: 97% CAS No.: 201733-56-4 Molecular Formula: C10H20B2O4 Formula Weight: 225.89 Storage Temperature: 2°C to 8°C Melting Point: 170-178° Beilstein Reference: 8839038 MDL No.: MFCD02093062

Formula: C₁₀H₂₀B₂O₄
MW: 225.89 g/mol
Melting Pt: 170…178 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093062
CAS Number: 201733-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR