4-Bromo-2-fluorobenzyl alcohol ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-Bromo-2-fluorobenzyl Alcohol, (4-Bromo-2-fluorophenyl)methanol

L18679.06
ALFAL18679.06EA 170 SGD
ALFAL18679.06
4-Bromo-2-fluorobenzyl alcohol ≥97%
4-Bromo-2-fluorobenzyl alcohol

Purity: 97% CAS No.: 188582-62-9 Molecular Formula: C7H6BrFO Formula Weight: 205.03 Melting Point: 31-33° Beilstein Reference: 7757667

Formula: C₇H₆BrFO
Melting Pt: 31…33 °C
Storage Temperature: Ambient
MDL Number: MFCD00143265
CAS Number: 188582-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR