4-Iodobenzyl alcohol ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (4-Iodophenyl)methanol

L19278.03 L19278.06
ALFAL19278.03EA 105 SGD
ALFAL19278.03 ALFAL19278.06
4-Iodobenzyl alcohol ≥97%
4-Iodobenzyl alcohol

Purity: 97% CAS No.: 18282-51-4 Molecular Formula: C7H7IO Formula Weight: 234.04 Melting Point: 72-74° Beilstein Reference: 1931621

Formula: C₇H₇IO
Melting Pt: 72…74 °C
Storage Temperature: Ambient
MDL Number: MFCD01732720
CAS Number: 18282-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR