2'-Bromo-4'-fluoroacetophenone ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-(2-Bromo-4-fluorophenyl)ethanone

L19320.03 L19320.06
ALFAL19320.03EA 128 SGD
ALFAL19320.03 ALFAL19320.06
2'-Bromo-4'-fluoroacetophenone ≥98%
2'-Bromo-4'-fluoroacetophenone

Purity: 98% CAS No.: 1006-39-9 Molecular Formula: C8H6BrFO Formula Weight: 217.04 Flash Point: 110°(230°F) Refractive Index: 1.5490 Boiling Point: 150°/12mm Beilstein Reference: 2249797 MDL No.: MFCD00077464

Formula: C₈H₆BrFO
MW: 217.04 g/mol
Boiling Pt: 150 °C (12 mmHg)
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00077464
CAS Number: 1006-39-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR