S-Thiouronium chloride, polymer-supported 0,8 - 1,0 mmol/g on Merrifield resin
Supplier: Thermo Fisher Scientific
L19377.03
ALFAL19377.03EA 0 SGD
ALFAL19377.03
S-Thiouronium chloride, polymer-supported 0,8 - 1,0 mmol/g on Merrifield resin
S-Thiouronium chloride, polymer-supported
MDL No.: MFCD03458600