2-Hydroxyphenylboronic acid ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Hydroxybenzeneboronic acid, Dihydroxy(2-hydroxyphenyl)borane, (2-Hydroxyphenyl)metaboric acid

L19400.03 L19400.06
ALFAL19400.03EA 109 SGD
ALFAL19400.03 ALFAL19400.06
2-Hydroxyphenylboronic acid ≥97%
2-Hydroxyphenylboronic acid

Purity: 97% CAS No.: 89466-08-0 Molecular Formula: C6H7BO3 Formula Weight: 137.93 Melting Point: ca 185° Beilstein Reference: 2962938

Formula: C₆H₇BO₃
MW: 137.93 g/mol
Melting Pt: ∼185 °C
Storage Temperature: Ambient
MDL Number: MFCD01074581
CAS Number: 89466-08-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR