3-Bromo-2-nitropyridine ≥98%

Supplier: Thermo Fisher Scientific
Warning

L19523.06 L19523.03
ALFAL19523.06EA 477 SGD
ALFAL19523.06 ALFAL19523.03
3-Bromo-2-nitropyridine ≥98%
3-Bromo-2-nitropyridine

Purity: 98+% CAS No.: 54231-33-3 Molecular Formula: C5H3BrN2O2 Formula Weight: 203.00 Melting Point: 102-106° Beilstein Reference: 1637071

Formula: C₅H₃BrN₂O₂
Melting Pt: 102…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00955614
CAS Number: 54231-33-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR