2-Trimethylsilyl-1-ethylboronic acid pinacol ester ≥97%

Supplier: Thermo Fisher Scientific

L19534.03 L19534.MD
ALFAL19534.03EA 0 SGD
ALFAL19534.03 ALFAL19534.MD
2-Trimethylsilyl-1-ethylboronic acid pinacol ester ≥97%
2-Trimethylsilyl-1-ethylboronic acid pinacol ester

Purity: 97% CAS No.: 165904-20-1 Molecular Formula: C11H25BO2Si Formula Weight: 228.22 Refractive Index: 1.4310 Beilstein Reference: 8683473

Formula: C₁₁H₂₅BO₂Si
MW: 228.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03788725
CAS Number: 165904-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR