5-Bromo-2-fluoropyridine ≥98%

Supplier: Thermo Fisher Scientific
Danger

L19537.06 L19537.14
ALFAL19537.06EA 129 SGD
ALFAL19537.06 ALFAL19537.14
5-Bromo-2-fluoropyridine ≥98%
5-Bromo-2-fluoropyridine

Purity: 98% CAS No.: 766-11-0 Molecular Formula: C5H3BrFN Formula Weight: 175.99 Flash Point: 73°(163°F) Refractive Index: 1.5294 Odour: Characteristic Boiling Point: 162-164° Beilstein Reference: 1363171 UN No.: 2810 MDL No.: MFCD01863742

Formula: C₅H₃BrFN
MW: 175.99 g/mol
Boiling Pt: 162…164 °C
Density: 1.72
Flash Pt: 73 °C (163 °F)
Storage Temperature: Ambient
MDL Number: MFCD01863742
CAS Number: 766-11-0
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR