2-(9-Carbazolyl)ethylboronic acid diethanolamine ester 98%

Supplier: Thermo Fisher Scientific

L19575.MC L19575.03
ALFAL19575.MCEA 0 SGD
ALFAL19575.MC ALFAL19575.03
2-(9-Carbazolyl)ethylboronic acid diethanolamine ester 98%
2-(9-Carbazolyl)ethylboronic acid diethanolamine ester

Purity: 98% CAS No.: 501014-45-5 Molecular Formula: C18H21BN2O2 Formula Weight: 308.19 Melting Point: 244-248° MDL No.: MFCD03788733

Formula: C₁₈H₂₁BN₂O₂
MW: 308.19 g/mol
Melting Pt: 244…248 °C
Storage Temperature: Ambient
MDL Number: MFCD03788733
CAS Number: 501014-45-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR