2-(9-Carbazolyl)ethylboronic acid pinacol ester ≥98%

Supplier: Thermo Fisher Scientific

Synonyms: 9-(2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)ethyl)-9H-carbazole

L19580.03 L19580.MD
ALFAL19580.03EA 482 SGD
ALFAL19580.03 ALFAL19580.MD
2-(9-Carbazolyl)ethylboronic acid pinacol ester ≥98%
2-(9-Carbazolyl)ethylboronic acid pinacol ester

Purity: 98% CAS No.: 608534-41-4 Molecular Formula: C20H24BNO2 Formula Weight: 321.23 Melting Point: 111-113° MDL No.: MFCD03788738

Formula: C₂₀H₂₄BNO₂
MW: 321.23 g/mol
Melting Pt: 111…113 °C
Storage Temperature: Ambient
MDL Number: MFCD03788738
CAS Number: 608534-41-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR