Phenol ∼3,5 mmol/g on polystyrene

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Phenylalcohol, Carbolic acid, Hydroxybenzene

L19594.06
ALFAL19594.06EA 0 SGD
ALFAL19594.06
Phenol ∼3,5 mmol/g on polystyrene
Phenol

Odour: Odourless MDL No.: MFCD03703209

Formula: C₆H₅OH
MW: 94.11 g/mol
Boiling Pt: 181.7
Melting Pt: 40.8
Flash Pt: 81
MDL Number: MFCD00002143
CAS Number: 108-95-2
Merck Index: 12,07390

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR