5-Bromo-2-iodopyridine ≥98%

Supplier: Thermo Fisher Scientific
Warning

L19610.14 L19610.06
ALFAL19610.14EA 292 SGD
ALFAL19610.14 ALFAL19610.06
5-Bromo-2-iodopyridine ≥98%
5-Bromo-2-iodopyridine

Purity: 98% CAS No.: 223463-13-6 Molecular Formula: C5H3BrIN Formula Weight: 283.89 Melting Point: 113-116° Beilstein Reference: 2119

Formula: C₅H₃BrIN
MW: 283.89 g/mol
Melting Pt: 113…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00691506
CAS Number: 223463-13-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR