Benzothiazole-2-carbonyl chloride 95%

Supplier: Thermo Fisher Scientific

L19631.03 L19631.MD
ALFAL19631.03EA 382 SGD
ALFAL19631.03 ALFAL19631.MD
Benzothiazole-2-carbonyl chloride 95%
Benzothiazole-2-carbonyl chloride

Purity: 95% CAS No.: 67748-61-2 Molecular Formula: C8H4ClNOS Formula Weight: 197.64 Melting Point: 113-115° Beilstein Reference: 130990 UN No.: UN3261 MDL No.: MFCD03659697

Formula: C₈H₄ClNOS
MW: 197.64 g/mol
Melting Pt: 113…115 °C
Storage Temperature: Ambient
MDL Number: MFCD03659697
CAS Number: 67748-61-2
ADR: 8 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR